Merck
  • Synthesis and properties of novel unsymmetrical donor molecules containing p-acetoxy- or p-hydroxyphenyl units.
Synthesis and properties of novel unsymmetrical donor molecules containing p-acetoxy- or p-hydroxyphenyl units.
Molecules (Basel, Switzerland) (2007-11-01)
Lakhemici Kaboub, Slimane Fradj, Abdelkrim Gouasmia
ABSTRACT
We report the synthesis and properties of eight new tetrathiafulvalene (TTF) derivatives containing two different functionalities, prepared with the aim of obtaining stable organic materials. The four acetoxyphenyl- and four hydroxyphenyl TTFs were synthesized via Wittig-type condensations. The electrochemical properties of these redox-active molecules were studied by cyclic voltammetry. Charge transfer complexes with tetracyanoquinodimethane (TCNQ) were prepared by chemical redox reactions. The complexes have been proven to give conducting materials. The UV-VIS and IR spectra of the TCNQ salts were recorded and used to characterize and estimate the degree of charge transfer of these complexes.
Product Description
Supelco
1,1,2-Trichloroethane, analytical standard
Sigma-Aldrich
1,1,2-Trichloroethane, contains ≤3% 2-propanol as stabilizer, 97%