Merck
  • High-yield 5-keto-D-gluconic acid formation is mediated by soluble and membrane-bound gluconate-5-dehydrogenases of Gluconobacter oxydans.
High-yield 5-keto-D-gluconic acid formation is mediated by soluble and membrane-bound gluconate-5-dehydrogenases of Gluconobacter oxydans.
Applied microbiology and biotechnology (2006-07-06)
Marcel Merfort, Ute Herrmann, Stephanie Bringer-Meyer, Hermann Sahm
ABSTRACT
Gluconobacter oxydans DSM 2343 is known to catalyze the oxidation of glucose to gluconic acid, and subsequently, to 2-keto-D-gluconic acid (2-KGA) and 5-keto-D-gluconic acid (5-KGA), by membrane-bound and soluble dehydrogenases. In G. oxydans MF1, in which the membrane-bound gluconate-2-dehydrogenase complex was inactivated, formation of the undesired 2-KGA was absent. This mutant strain uniquely accumulates high amounts of 5-KGA in the culture medium. To increase the production rate of 5-KGA, which can be converted to industrially important L-(+)-tartaric acid, we equipped G. oxydans MF1 with plasmids allowing the overproduction of the soluble and the membrane-bound 5-KGA-forming enzyme. Whereas the overproduction of the soluble gluconate:NADP 5-oxidoreductase resulted in the accumulation of up to 200 mM 5-KGA, the detected 5-KGA accumulation was even higher when the gene coding for the membrane-bound gluconate-5-dehydrogenase was overexpressed (240 to 295 mM 5-KGA). These results provide a basis for designing a biotransformation process for the conversion of glucose to 5-KGA using the membrane-bound as well as the soluble enzyme system.
Sigma-Aldrich
5-Keto-D-gluconic acid potassium salt, ≥98.0% (TLC)
Pricing and availability is not currently available.