Merck
  • Concerted proton motion in hydrogen-bonded trimers: a spontaneous Raman scattering perspective.
Concerted proton motion in hydrogen-bonded trimers: a spontaneous Raman scattering perspective.
Physical chemistry chemical physics : PCCP (2006-06-16)
Philipp Zielke, Martin A Suhm
ABSTRACT
Raman active symmetric O-H stretching modes are detected and assigned for the first time in isolated methanol, ethanol and methyl lactate trimers, providing insights into cluster structure, vibrational assignments and hydrogen-bond mediated couplings.
Sigma-Aldrich
Methyl DL-lactate, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Methyl D-lactate, 98%, optical purity ee: 96% (GLC)
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Methyl L-lactate, 98%, optical purity ee: 97% (GLC)
Pricing and availability is not currently available.