- Impact of the gliotoxin L-serine-O-sulphate on cellular metabolism in cultured rat astrocytes.
Impact of the gliotoxin L-serine-O-sulphate on cellular metabolism in cultured rat astrocytes.
L-serine-O-sulphate is a member of a group of amino acids collectively called gliotoxins and is a substrate for the high affinity sodium-dependent glutamate transporters. Previous studies have shown that it is toxic to primary cultures of astrocytes but the mode of toxicity is unknown. The current study demonstrates that L-serine-O-sulphate, at a sub-toxic concentration (400 microM), causes significant disruption to glucose and alanine metabolism in cultures of rat cortical astrocytes. More specifically, using (13)C NMR spectroscopy a significant reduction in labelled end products from [1-(13)C]glucose and [3-(13)C]alanine was found in the presence of L-serine-O-sulphate. Additionally, using [2-(13)C]glycine a 27% reduction in de novo glutathione synthesis was observed in the presence of the gliotoxin. Incubation of the cells with L-serine-O-sulphate reduced the activity of alanine and aspartate aminotransferase by 53% and 67%, respectively. Collectively these results show that the gliotoxin, L-serine-O-sulphate, causes major disruptions to metabolic pathways in primary cultures of astrocytes.