Merck
  • G-protein-coupled receptor chromatographic stationary phases. 2. Ligand-induced conformational mobility in an immobilized beta2-adrenergic receptor.
G-protein-coupled receptor chromatographic stationary phases. 2. Ligand-induced conformational mobility in an immobilized beta2-adrenergic receptor.
Analytical chemistry (2004-12-15)
Farideh Beigi, Khalid Chakir, Rui-Ping Xiao, Irving W Wainer
ABSTRACT
Membranes from a HEK-293 cell line expressing the beta(2)-adrenergic receptor (beta(2)-AR) have been immobilized on an artificial membrane liquid chromatographic stationary phase. The resulting phase was packed into a glass column (1.8 x 0.5 (i.d.) cm) and used in on-line chromatographic system. Frontal displacement affinity chromatography was used to determine the dissociation constants (K(d)) of CGP 12177A (552.6 nM) and (S)-propranolol (84.3 nM). Zonal displacement chromatography using CGP 12177A as the marker and racemic mixtures of the antagonists nadolol and propranolol demonstrated that the immobilized beta(2)-AR retained its ability to specifically bind these compounds. Similar experiments with (R)- and (S)-propranolol demonstrated that the immobilized receptor retained its enantioselectivity as (S)-propranolol displaced the CGP 12177 marker to a great extent that the (R)-enantiomer. The addition of the agonist butoxamine to the mobile phase increased the retention of the CGP-12177A as did the addition of the agonist fenoterol. These results indicate that the immobilized beta(2)-AR retained its ability to undergo ligand-induced conformational changes. The data from this study suggest that the immobilized beta(2)-AR can be used to screen for ligand binding interactions in both the resting and active states of the receptor.
Sigma-Aldrich
(±)-CGP-12177A, ≥98% (HPLC), solid
Supelco
Butoxamine hydrochloride, analytical standard, for drug analysis, mixture of diastereomers