Merck
  • GDNF: a glial cell line-derived neurotrophic factor for midbrain dopaminergic neurons.
GDNF: a glial cell line-derived neurotrophic factor for midbrain dopaminergic neurons.
Science (New York, N.Y.) (1993-05-21)
L F Lin, D H Doherty, J D Lile, S Bektesh, F Collins
ABSTRACT
A potent neurotrophic factor that enhances survival of midbrain dopaminergic neurons was purified and cloned. Glial cell line-derived neurotrophic factor (GDNF) is a glycosylated, disulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfamily. In embryonic midbrain cultures, recombinant human GDNF promoted the survival and morphological differentiation of dopaminergic neurons and increased their high-affinity dopamine uptake. These effects were relatively specific; GDNF did not increase total neuron or astrocyte numbers nor did it increase transmitter uptake by gamma-aminobutyric-containing and serotonergic neurons. GDNF may have utility in the treatment of Parkinson's disease, which is marked by progressive degeneration of midbrain dopaminergic neurons.
Sigma-Aldrich
GDNF human, recombinant, expressed in E. coli, Animal-component free, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
GDNF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
GDNF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture