Merck
  • Stereospecificity in the Au-catalysed cyclisation of monoallylic diols. Synthesis of (+)-isoaltholactone.
Stereospecificity in the Au-catalysed cyclisation of monoallylic diols. Synthesis of (+)-isoaltholactone.
Chemical communications (Cambridge, England) (2011-06-15)
William P Unsworth, Kiri Stevens, Scott G Lamont, Jeremy Robertson
ABSTRACT
We describe a concise synthesis of (+)-isoaltholactone via a Au-catalysed cyclisation of a monoallylic diol to form the tetrahydrofuranyl ring. Analogous cyclisations show that the stereochemical outcome is dictated by the stereochemistry of the diol substrate.
Sigma-Aldrich
Altholactone, ≥95% (LC/MS-ELSD)