Merck
Cloning and characterization of human pancreatic lipase cDNA.
The Journal of biological chemistry (1989-11-25)
M E Lowe, J L Rosenblum, A W Strauss
ABSTRACT
Pancreatic lipase (triacylglycerol acylhydrolase, EC 3.1.1.3) hydrolyzes dietary long chain triacylglycerol to free fatty acids and monoacylglycerols in the intestinal lumen. In the presence of bile acids, the activity of lipase is stimulated by colipase. As a prelude to studying the relationship of the protein structures to the functional properties of lipase and colipase, a cDNA encoding human pancreatic lipase was isolated from a lambda gt11 cDNA library screened with a rabbit polyclonal anti-human pancreatic lipase antibody. The full length cDNA clone of 1477 base pairs contained an open reading frame encoding a 465-amino acid protein, including a 16-amino acid signal peptide. The nucleotide sequence was 69% identical to the dog pancreatic lipase cDNA. The predicted NH2-terminal protein sequence agreed with the published NH2-terminal sequence of human pancreatic lipase and the predicted protein sequence was 85 and 70% identical to the protein sequences of pig and dog pancreatic lipase, respectively. A region of homology around Ser-153 is conserved in a number of lipid-binding proteins. Human hepatic lipase and lipoprotein lipase share extensive homology with pancreatic lipase, suggesting that the three proteins are members of a small gene family. In vitro translation of mRNA transcribed from the cDNA resulted in a protein of the expected molecular size that could be processed by microsomal membranes to yield a glycolated protein with proper signal peptide cleavage. RNA blot analysis demonstrated tissue specificity for pancreatic lipase. Thus, for the first time, a full length human pancreatic lipase cDNA has been isolated and characterized. The demonstrated regions of homology with other lipases will aid definition of interactions with substrate and colipase through site-specific mutagenesis.
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg