- NMR characterization of the interaction between the PUB domain of peptide:N-glycanase and ubiquitin-like domain of HR23.
NMR characterization of the interaction between the PUB domain of peptide:N-glycanase and ubiquitin-like domain of HR23.
FEBS letters (2012-05-12)
Yukiko Kamiya, Yoshinori Uekusa, Akira Sumiyoshi, Hiroaki Sasakawa, Takeshi Hirao, Tadashi Suzuki, Koichi Kato
PMID22575648
ABSTRACT
PUB domains are identified in several proteins functioning in the ubiquitin (Ub)-proteasome system and considered as p97-binding modules. To address the further functional roles of these domains, we herein characterized the interactions of the PUB domain of peptide:N-glycanase (PNGase) with Ub and Ub-like domain (UBL) of the proteasome shuttle factor HR23. NMR data indicated that PNGase-PUB exerts an acceptor preferentially for HR23-UBL, electrostatically interacting with the UBL surface employed for binding to other Ub/UBL motifs. Our findings imply that PNGase-PUB serves not only as p97-binding module but also as a possible activator of HR23 in endoplasmic reticulum-associated degradation mechanisms.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE), for proteomics
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit