Merck
  • Adenosine transporter ENT1 regulates the acquisition of goal-directed behavior and ethanol drinking through A2A receptor in the dorsomedial striatum.
Adenosine transporter ENT1 regulates the acquisition of goal-directed behavior and ethanol drinking through A2A receptor in the dorsomedial striatum.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-03-08)
Hyung Wook Nam, David J Hinton, Na Young Kang, Taehyun Kim, Moonnoh R Lee, Alfredo Oliveros, Chelsea Adams, Christina L Ruby, Doo-Sup Choi
ABSTRACT
Adenosine signaling has been implicated in the pathophysiology of many psychiatric disorders including alcoholism. Striatal adenosine A2A receptors (A2AR) play an essential role in both ethanol drinking and the shift from goal-directed action to habitual behavior. However, direct evidence for a role of striatal A2AR signaling in ethanol drinking and habit development has not been established. In the present study, we found that decreased A2AR-mediated CREB activity in the dorsomedial striatum (DMS) enhanced initial behavioral acquisition of goal-directed behaviors and the vulnerability to progress to excessive ethanol drinking during operant conditioning in mice lacking ethanol-sensitive adenosine transporter ENT1 (ENT1(-/-)). Using mice expressing β-galactosidase (lacZ) under the control of seven repeated CRE sites in both genotypes (CRE-lacZ/ENT1(+/+) mice and CRE-lacZ/ENT1(-/-) mice) and the dominant-negative form of CREB, we found that reduced CREB activity in the DMS was causally associated with decreased A2AR signaling and increased goal-directed ethanol drinking. Finally, we have demonstrated that the A2AR antagonist ZM241385 dampened protein kinase A activity-mediated signaling in the DMS and promoted excessive ethanol drinking in ENT1(+/+) mice, but not in ENT1(-/-) mice. Our results indicate that A2AR-mediated CREB signaling in the DMS is a key determinant in enhancing the development of goal-directed ethanol drinking in mice.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse