- [Effect of combined therapy with melaxen on lipid peroxidation rate and citrate content in blood serum of patients with drug-induced hepatitis diagnosis].
[Effect of combined therapy with melaxen on lipid peroxidation rate and citrate content in blood serum of patients with drug-induced hepatitis diagnosis].
Eksperimental'naia i klinicheskaia farmakologiia (2012-10-03)
S S Popov, K K Shul'gin, A N Pashkov, V I Zoloedov, G I Shvedov
PMID23025050
ABSTRACT
A combined therapy with melaxen led to a decrease in the activity of gamma-glutamyl transpeptidase and the level of diene conjugates in blood serum of patients with the drug-induced hepatitis developing on the background of administration of antituberculous preparations. These changes are indicative of pronounced antioxidant and hepatoprotective properties of the drug. In addition, there was a decrease in the content of citrate and a change in the activity of aconitate hydratase toward a normal level, which reflects a decrease in the degree of pathologic oxidative stress development and is evidence of the antiradical effect of melaxen.
MATERIALS