- Clostridium histolyticum collagenase in the treatment of Dupuytren's contracture.
Clostridium histolyticum collagenase in the treatment of Dupuytren's contracture.
British journal of hospital medicine (London, England : 2005) (2012-08-10)
Ernest Azzopardi, Dean E Boyce
PMID22875519
ABSTRACT
Dupuytren's disease is a common, costly and recurrent health issue. This review compares Clostridium histolyticum collagenase with current operative treatments. Collagenase management is an effective non-surgical alternative associated with lower risks of serious adverse events, but higher incidence of non-serious adverse events.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)