Merck
  • Microbial isoprenoid biosynthesis and human gammadelta T cell activation.
Microbial isoprenoid biosynthesis and human gammadelta T cell activation.
FEBS letters (2003-06-05)
Matthias Eberl, Martin Hintz, Armin Reichenberg, Ann-Kristin Kollas, Jochen Wiesner, Hassan Jomaa
ABSTRACT
Human Vgamma9/Vdelta2 T cells play a crucial role in the immune response to microbial pathogens, yet their unconventional reactivity towards non-peptide antigens has been enigmatic until recently. The break-through in identification of the specific activator was only possible due to recent success in a seemingly remote field: the elucidation of the reaction steps of the newly discovered 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway of isoprenoid biosynthesis that is utilised by many pathogenic bacteria. Unexpectedly, the intermediate of the MEP pathway, (E)-4-hydroxy-3-methyl-but-2-enyl-pyrophosphate) (HMB-PP), turned out to be by far the most potent Vgamma9/Vdelta2 T cell activator known, with an EC(50) of 0.1 nM.
Sigma-Aldrich
(E)-1-Hydroxy-2-methyl-2-butenyl 4-pyrophosphate lithium salt, ≥95.0% (TLC)