Merck
  • Functional and proliferative effects of repeated low-dose oral administration of furan in rat liver.
Functional and proliferative effects of repeated low-dose oral administration of furan in rat liver.
Molecular nutrition & food research (2010-06-12)
Angela Mally, Carmen Graff, Olga Schmal, Sabrina Moro, Carolin Hamberger, Ute M Schauer, Jens Brück, Sibel Ozden, Max Sieber, Ulrich Steger, Dieter Schrenk, Gordon C Hard, James Kevin Chipman, Wolfgang Dekant
ABSTRACT
Furan, a food contaminant formed during heat processing, induces hepatocellular tumors in rodents and high incidences of cholangiocarcinomas in rats even at the lowest dose (2 mg/kg b.w.) administered. Initial estimates suggested that human intake of furan may be as high as 3.5 μg/kg b.w./day, indicating a relatively narrow margin of exposure. The aim of this study was to establish dose-response data for cytotoxicity, regenerative cell proliferation and secondary oxidative DNA damage in livers of male F344 rats treated with furan at doses ≤2 mg/kg b.w. for 28 days. No significant signs of hepatotoxicity other than a mild, dose-dependent increase in serum cholesterol and unconjugated bile acids, and no evidence of oxidative DNA damage were seen. Histopathological alterations and proliferative changes were restricted to subcapsular areas of the left and caudate liver lobes. Although statistically significant effects were only seen at the 2 mg/kg b.w. dose during the course of our study, a ∼two and ∼threefold increase in 5-bromo-2'-deoxyuridine labeling index was observed at 0.1 and 0.5 mg/kg b.w., respectively, suggesting that chronic exposure to doses even below 2 mg/kg b.w. may cause proliferative changes in rat liver and highlighting the need to assess furan carcinogenicity at lower doses.
Sigma-Aldrich
Sodium taurohyodeoxycholate hydrate, ≥98%