Merck
Association of CD2 and CD45 on human T lymphocytes.
Nature (1990-05-03)
B Schraven, Y Samstag, P Altevogt, S C Meuer
ABSTRACT
At least two membrane receptors have been defined through which human T lymphocytes can be induced to proliferate and differentiate, namely the CD3-Ti antigen receptor complex and the CD2 molecule. Monoclonal antibodies directed at either CD2 or CD3 induce intracellular second messenger production and subsequent protein phosphorylation. On most human non-B lymphocytes, CD3-Ti and CD2 are coexpressed and seem to be functionally interrelated. But there are minor subpopulations in which these receptor systems can transduce signals despite a mutually exclusive expression, indicating that CD3-Ti and CD2 can act independently of each other. This view is supported by the finding that most monoclonal antibodies directed at the CD45 molecules are strongly co-mitogenic with CD2 but not CD3 monoclonal antibodies. As the intracytoplasmic domains of CD45 have tyrosine phosphatase activity these functional effects could be explained by a physical association between CD2 and CD45. Using chemical crosslinking techniques, we now show that CD45 is linked to CD2 on the surface of human T lymphocytes.
Sigma-Aldrich
CD2 human, recombinant, expressed in E. coli, 0.5 mg protein/mL
Pricing and availability is not currently available.