Merck
  • Sestrin 2 Deficiency Exacerbates Noise-Induced Cochlear Injury Through Inhibiting ULK1/Parkin-Mediated Mitophagy.
Sestrin 2 Deficiency Exacerbates Noise-Induced Cochlear Injury Through Inhibiting ULK1/Parkin-Mediated Mitophagy.
Antioxidants & redox signaling (2022-06-17)
Yalan Li, Shengsheng Li, Liyuan Wu, Tingting Wu, Mengxiao Li, Deliang Du, Yalin Chen, Caiji Wang, Xuanyi Li, Shili Zhang, Zeqi Zhao, Liting Zheng, Mengbing Chen, Menghua Li, Ting Li, Xi Shi, Yuehua Qiao
ABSTRACT
Aims: Noise damage to auditory hair cells is associated with oxidative stress and mitochondrial dysfunction. This study aimed to investigate the possible effect of sestrin 2 (SESN2), an endogenous antioxidant protein, on noise-induced hearing loss (NIHL) and the underlying mechanisms. Results: We identified SESN2 as a protective factor against oxidative stress in NIHL through activation of Parkin-mediated mitophagy. Consistently, SESN2 expression was increased and mitophagy was induced during the early stage after a temporary threshold shift due to noise exposure or hydrogen peroxide(H2O2) stimulation; conversely, SESN2 deficiency blocked mitophagy and exacerbated acoustic trauma. Mechanistically, SESN2 interacted with Unc-51-like protein kinase 1(ULK1), promoting ULK1 protein-level stabilization by interfering with its proteasomal degradation. This stabilization is essential for mitophagy initiation, since restoring ULK1 expression in SESN2-silenced cells rescued mitophagy defects. Innovation and Conclusion: Our results provide novel insights regarding SESN2 as a therapeutic target against noise-induced cochlear injury, possibly through improved mitophagy. Antioxid. Redox Signal. 38, 115-136.
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution