Merck
  • The RNA binding proteins LARP4A and LARP4B promote sarcoma and carcinoma growth and metastasis.
The RNA binding proteins LARP4A and LARP4B promote sarcoma and carcinoma growth and metastasis.
iScience (2024-03-27)
Jennifer C Coleman, Luke Tattersall, Val Yianni, Laura Knight, Hongqiang Yu, Sadie R Hallett, Philip Johnson, Ana J Caetano, Charlie Cosstick, Anne J Ridley, Alison Gartland, Maria R Conte, Agamemnon E Grigoriadis
ABSTRACT
RNA-binding proteins (RBPs) are emerging as important regulators of cancer pathogenesis. We reveal that the RBPs LARP4A and LARP4B are differentially overexpressed in osteosarcoma and osteosarcoma lung metastases, as well as in prostate cancer. Depletion of LARP4A and LARP4B reduced tumor growth and metastatic spread in xenografts, as well as inhibiting cell proliferation, motility, and migration. Transcriptomic profiling and high-content multiparametric analyses unveiled a central role for LARP4B, but not LARP4A, in regulating cell cycle progression in osteosarcoma and prostate cancer cells, potentially through modulating key cell cycle proteins such as Cyclins B1 and E2, Aurora B, and E2F1. This first systematic comparison between LARP4A and LARP4B assigns new pro-tumorigenic functions to LARP4A and LARP4B in bone and prostate cancer, highlighting their similarities while also indicating distinct functional differences. Uncovering clear biological roles for these paralogous proteins provides new avenues for identifying tissue-specific targets and potential druggable intervention.
Sigma-Aldrich
Anti-LARP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-LARP4B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution