Merck
  • A simple and robust nanosystem for photoacoustic imaging of bladder cancer based on α5β1-targeted gold nanorods.
A simple and robust nanosystem for photoacoustic imaging of bladder cancer based on α5β1-targeted gold nanorods.
Journal of nanobiotechnology (2023-08-28)
Massimo Alfano, Elisa Alchera, Angelina Sacchi, Alessandro Gori, Giacomo Quilici, Irene Locatelli, Chiara Venegoni, Roberta Lucianò, Anna Maria Gasparri, Barbara Colombo, Giulia Taiè, Jithin Jose, Paolo Armanetti, Luca Menichetti, Giovanna Musco, Andrea Salonia, Angelo Corti, Flavio Curnis
ABSTRACT
Early detection and removal of bladder cancer in patients is crucial to prevent tumor recurrence and progression. Because current imaging techniques may fail to detect small lesions of in situ carcinomas, patients with bladder cancer often relapse after initial diagnosis, thereby requiring frequent follow-up and treatments. In an attempt to obtain a sensitive and high-resolution imaging modality for bladder cancer, we have developed a photoacoustic imaging approach based on the use of PEGylated gold nanorods (GNRs) as a contrast agent, functionalized with the peptide cyclic [CphgisoDGRG] (Iso4), a selective ligand of α5β1 integrin expressed by bladder cancer cells. This product (called GNRs@PEG-Iso4) was produced by a simple two-step procedure based on GNRs activation with lipoic acid-polyethyleneglycol(PEG-5KDa)-maleimide and functionalization with peptide Iso4. Biochemical and biological studies showed that GNRs@PEG-Iso4 can efficiently recognize purified integrin α5β1 and α5β1-positive bladder cancer cells. GNRs@PEG-Iso4 was stable and did not aggregate in urine or in 5% sodium chloride, or after freeze/thaw cycles or prolonged exposure to 55 °C, and, even more importantly, do not settle after instillation into the bladder. Intravesical instillation of GNRs@PEG-Iso4 into mice bearing orthotopic MB49-Luc bladder tumors, followed by photoacoustic imaging, efficiently detected small cancer lesions. The binding to tumor lesions was competed by a neutralizing anti-α5β1 integrin antibody; furthermore, no binding was observed to healthy bladders (α5β1-negative), pointing to a specific targeting mechanism. GNRs@PEG-Iso4 represents a simple and robust contrast agent for photoacoustic imaging and diagnosis of small bladder cancer lesions.
Sigma-Aldrich
Anti-Rat IgM (μ-chain specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
IgG1 Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Integrin αVβ6 Antibody, clone 10D5, azide free, clone 10D5, Chemicon®, from mouse