- Inhibition of TERC inhibits neural apoptosis and inflammation in spinal cord injury through Akt activation and p-38 inhibition via the miR-34a-5p/XBP-1 axis.
Inhibition of TERC inhibits neural apoptosis and inflammation in spinal cord injury through Akt activation and p-38 inhibition via the miR-34a-5p/XBP-1 axis.
This study investigated the function of telomerase RNA component (TERC) in spinal cord injury (SCI). SCI models were established in rats via laminectomy and PC-12 cells were treated with lipopolysaccharide (LPS). TERC and miR-34a-5p expressions in cells and rat spinal cords were detected by quantitative reverse transcription polymerase chain reaction, followed by overexpression/knockdown of TERC/miR-34a-5p. Spinal cord histopathological changes were examined via hematoxylin-eosin staining. miR-34a-5p' relation with TERC and XBP-1 was predicted by TargetScan and checked by dual-luciferase reporter/RNA immunoprecipitation assays. Cell biological behaviors were assessed by Cell counting kit-8, wound healing, Transwell, and flow cytometry assays. XBP-1 and inflammation/apoptosis-related protein expressions were analyzed by western blot. TERC was upregulated and miR-34a-5p was low-expressed in SCI tissues and LPS-induced PC-12 cells. TERC-knockdown alleviated histopathological abnormalities yet upregulated miR-34a-5p in SCI tissues. In LPS-induced PC-12 cells, TERC knockdown promoted cell viability, migration, invasion, and inhibited apoptosis, while TERC overexpression ran oppositely. TERC knockdown downregulated the XBP-1, IL-6, TNF-α, Bax, p-p38/t-p38, and cleaved caspase-9/-3, but upregulated Bcl-2 and p-Akt/t-Akt. TERC targeted miR-34a-5p, which further targeted XBP-1. miR-34a-5p downregulation exerted effects opposite to and offset TERC knockdown-induced effects. TERC knockdown facilitated the regeneration of neuron tissues yet inhibited inflammation in SCI through Akt activation and p-38 inhibition via the miR-34a-5p/XBP-1 axis.