Merck
  • SUN11602-induced hyperexpression of calbindin D-28k is pivotal for the survival of hippocampal neurons under neurotoxic conditions.
SUN11602-induced hyperexpression of calbindin D-28k is pivotal for the survival of hippocampal neurons under neurotoxic conditions.
Brain research (2014-12-03)
Norihito Murayama, Takafumi Noshita, Ryoko Ogino, Toyofumi Masuda, Taisuke Kadoshima, Tetsushi Oka, Nobuhiro Ueno, Naohiro Takemoto, Tetsuya Toba, Shinya Ueno, Wiebke Schulze, Yoshiyuki Igawa, Yasuhiro Morita, Teruyoshi Inoue
ABSTRACT
Basic fibroblast growth factor (FGF-2/bFGF) possesses neuroprotective activity and promotes cell proliferation. In this study, the novel synthetic compound 4-({4-[[(4-amino-2,3,5,6-tetramethylanilino)acetyl](methyl)amino]-1-piperidinyl}methyl)benzamide (SUN11602) exhibited neuroprotective activities similar to those of FGF-2 without promoting cell proliferation. In primary cultures of hippocampal neurons, stimulation with SUN11602 or FGF-2 increased calbindin D-28k (CalB) gene expression and prevented glutamate-induced neuronal death. These effects were abolished by pretreatment with PD166866 (FGF receptor 1 [FGFR1] tyrosine kinase-specific inhibitor). This indicated that FGFR1 activation and increased CalB expression were involved in SUN11602-mediated neuroprotection. However, receptor-binding assays revealed that unlike FGF-2, SUN11602 did not alter the binding of (125)I-labeled FGF-2 to FGFR1. To investigate the possible proliferative activity of SUN11602, we utilized BHK21 and SKN cells expressing endogenous FGFR1. FGF-2 promoted cell proliferation whereas SUN11602 did not. In in vivo studies, wild-type (WT) and CalB-deficient (CalB(-/-)) mice were injected with aggregated Aβ1-40 and ibotenate (NMDA receptor agonist) to severely damage the hippocampal tissue. Treatment with SUN11602 (orally) or FGF-2 (intraparenchymally) at the midpoint of Aβ1-40 and ibotenate injections prevented the hippocampal damage in WT mice, however this effect was abolished in CalB(-/-) mice. Thus, SUN11602 exerted protective effects on hippocampal neurons through activation of FGFR1 and increased CalB expression. Moreover, the neuroprotective effects of SUN11602 depended upon the various biological activities of FGF-2.
Sigma-Aldrich
SUN11602, ≥95% (HPLC)