Merck
  • THOC5 complexes with DDX5, DDX17, and CDK12 to regulate R loop structures and transcription elongation rate.
THOC5 complexes with DDX5, DDX17, and CDK12 to regulate R loop structures and transcription elongation rate.
iScience (2023-01-03)
Mareike Polenkowski, Aldrige Bernardus Allister, Sebastian Burbano de Lara, Andrew Pierce, Bethany Geary, Omar El Bounkari, Lutz Wiehlmann, Andrea Hoffmann, Anthony D Whetton, Teruko Tamura, Doan Duy Hai Tran
ABSTRACT
THOC5, a member of the THO complex, is essential for the 3'processing of some inducible genes, the export of a subset of mRNAs and stem cell survival. Here we show that THOC5 depletion results in altered 3'cleavage of >50% of mRNAs and changes in RNA polymerase II binding across genes. THOC5 is recruited close to high-density polymerase II sites, suggesting that THOC5 is involved in transcriptional elongation. Indeed, measurement of elongation rates in vivo demonstrated decreased rates in THOC5-depleted cells. Furthermore, THOC5 is preferentially recruited to its target genes in slow polymerase II cells compared with fast polymerase II cells. Importantly chromatin-associated THOC5 interacts with CDK12 (a modulator of transcription elongation) and RNA helicases DDX5, DDX17, and THOC6 only in slow polymerase II cells. The CDK12/THOC5 interaction promotes CDK12 recruitment to R-loops in a THOC6-dependent manner. These data demonstrate a novel function of THOC5 in transcription elongation.
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Anti-THOC7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2