Merck
  • Targeted elimination of mutated mitochondrial DNA by a multi-functional conjugate capable of sequence-specific adenine alkylation.
Targeted elimination of mutated mitochondrial DNA by a multi-functional conjugate capable of sequence-specific adenine alkylation.
Cell chemical biology (2021-08-28)
Takuya Hidaka, Kaori Hashiya, Toshikazu Bando, Ganesh N Pandian, Hiroshi Sugiyama
ABSTRACT
Mutations in mitochondrial DNA (mtDNA) cause mitochondrial diseases, characterized by abnormal mitochondrial function. Although eliminating mutated mtDNA has potential to cure mitochondrial diseases, no chemical-based drugs in clinical trials are capable of selective modulation of mtDNA mutations. Here, we construct a class of compounds encompassing pyrrole-imidazole polyamides (PIPs), mitochondria-penetrating peptide, and chlorambucil, an adenine-specific DNA-alkylating reagent. The sequence-selective DNA binding of PIPs allows chlorambucil to alkylate mutant adenine more efficiently than other sites in mtDNA. In vitro DNA alkylation assay shows that our compound 8950A-Chb(Cl/OH) targeting a nonpathogenic point mutation in HeLa S3 cells (m.8950G>A) can specifically alkylate the mutant adenine. Furthermore, the compound reduces the mtDNA possessing the target mutation in cultured HeLa S3 cells. The programmability of PIPs to target different sequences could allow this class of compounds to be developed as designer drugs targeting pathogenic mutations associated with mitochondrial diseases in future studies.
Sigma-Aldrich
Chlorambucil