- Comparison of specific gravity and creatinine for normalizing urinary reproductive hormone concentrations.
Comparison of specific gravity and creatinine for normalizing urinary reproductive hormone concentrations.
Specific gravity (SG) may perform as well as creatinine (CR) correction for adjusting urinary hormone concentrations, as well as offer some advantages. We compared the two methods and applied them to US and Bangladeshi specimens to evaluate their use in different populations. Pearson correlations between serum concentrations and SG, CR, and uncorrected urinary concentrations were compared using paired daily urine and serum specimens from one menstrual cycle from 30 US women. Corrected urinary estrone conjugate and pregnanediol glucuronide concentrations were compared with serum estradiol and progesterone. Urine specimens across one menstrual cycle from 13 Bangladeshi women were used to evaluate the applicability of both methods to a nonindustrialized population. Linear mixed-effects models were used to compare CR and SG values in the Bangladeshi vs US specimens. There was no significant difference between SG-corrected vs serum and CR-corrected vs serum correlations for either assay. Usable CR results were obtained for all US specimens, but 37% of the Bangladeshi specimens were below the CR assay limit of detection. The Bangladeshi sample had significantly lower CR and higher inter- and intrasubject CR variability than the US sample. SG is a potentially useful alternative to CR correction for normalizing urinary steroid hormone concentrations, particularly in settings where CR values are highly variable or unusually low.