Merck
  • 3-Amino-fluorene-2,4-dicarbonitriles (AFDCs) as Photocatalysts for the Decarboxylative Arylation of α-Amino Acids and α-Oxy Acids with Arylnitriles.
3-Amino-fluorene-2,4-dicarbonitriles (AFDCs) as Photocatalysts for the Decarboxylative Arylation of α-Amino Acids and α-Oxy Acids with Arylnitriles.
Organic letters (2019-03-13)
Yiyang Chen, Ping Lu, Yanguang Wang
ABSTRACT
1-(4-(9 H-Carbazol-9-yl)phenyl)-3-amino-9 H-fluorene-2,4-dicarbonitrile as a new photocatalyst for the decarboxylative cross-coupling reaction of α-amino acids or α-oxy carboxylic acids with arylnitriles is described. This light-driven reaction enables a variety of benzylic amines and ethers to be prepared from readily available starting materials under mild conditions.
Sigma-Aldrich
(±)-α-Methoxyphenylacetic acid, crystalline