Merck
  • Absence of gelatinase (MMP-9) or collagenase (MMP-13) attenuates adriamycin-induced albuminuria and glomerulosclerosis.
Absence of gelatinase (MMP-9) or collagenase (MMP-13) attenuates adriamycin-induced albuminuria and glomerulosclerosis.
Nephron. Experimental nephrology (2010-04-22)
Yusuke Sakamaki, Hiroyuki Sasamura, Kaori Hayashi, Kimiko Ishiguro, Hironari Takaishi, Yasunori Okada, Jeanine M D'Armiento, Takao Saruta, Hiroshi Itoh
ABSTRACT
The role of matrix metalloproteinases (MMPs) in the pathogenesis of glomerular injury appears to be complex. To investigate the role of individual MMPs, we examined the course of Adriamycin-induced albuminuria and glomerulosclerosis in mice lacking either a gelatinase (MMP-9) or a collagenase (MMP-13). Adriamycin was administered to MMP-9 or MMP-13 knockout (KO) mice. Glomerular injury was assessed by the quantification of albuminuria, the glomerular injury score and type IV collagen immunostaining. Treatment of mice with Adriamycin (18 mg/kg i.v.) resulted in marked albuminuria and glomerulosclerosis reaching a peak at 4-8 weeks. The albuminuria and glomerulosclerosis were significantly (p < 0.05) attenuated in both the MMP-9 KO and MMP-13 KO mice compared to controls. In contrast, treatment of wild-type mice with the broad-spectrum MMP inhibitor doxycycline did not have a beneficial effect on the albuminuria and glomerulosclerosis. These results support a role for both gelatinase (MMP-9) and collagenase (MMP-13) in the pathogenesis of glomerular injury in the Adriamycin-induced glomerulosclerosis model. MMP inhibitors with high specificity towards MMP-9 and/or MMP-13 may be potential future candidates to provide more effective therapies to inhibit the development of glomerulosclerosis.
Sigma-Aldrich
MMP-2 Inhibitor III, The MMP-2 Inhibitor III, also referenced under CAS 704888-90-4, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.