Merck
  • Resistance to fas-induced apoptosis in cells from human atherosclerotic lesions: elevated Bcl-XL inhibits apoptosis and caspase activation.
Resistance to fas-induced apoptosis in cells from human atherosclerotic lesions: elevated Bcl-XL inhibits apoptosis and caspase activation.
Journal of vascular research (2007-07-28)
Zhaoqing Yang, Dmitry Gagarin, Ali Ramezani, Robert G Hawley, Timothy A McCaffrey
ABSTRACT
The inappropriate survival of cells in the neointima contributes to atherosclerotic plaque progression, while apoptosis in the fibrous cap of lesions contributes to myocardial infarction and stroke. Prior genomic-scale transcript profiling of human carotid artery plaque cells with known sensitivity or resistance to fas-induced apoptosis identified candidate genes involved in lesion cell apoptosis. Retroviral overexpression indicated that several candidate factors were not causative, but that Bcl-X(L) conferred complete resistance to apoptosis induced by fas ligation. Resistant cells failed to efficiently activate caspase 8, an effect which was also observed in Bcl-X(L)-transfected cells. Small-molecule Bcl-2/X(L) inhibitors and siRNA knockdown of Bcl-X(L) markedly sensitized resistant cells to apoptosis, and partially restored caspase 8 activation. Caspase 3, 6 and 9 inhibitors reduced caspase 8 activation and blocked apoptosis. Complete knockdown of caspase 9 did not reduce apoptosis, while knockdown of Bid suppressed apoptosis, suggesting that mitochondrial pathways independent of caspase 9, such as Smac/Diablo or AIF, provide a necessary mitochondrial input to efficient caspase activation. Bcl-X(L) appears to modulate lesion cell apoptosis by suppressing mitochondrial amplification of caspase activation loops. The results may have direct implications for controlling plaque instability/progression, and identify a new class of small molecules to inhibit restenosis.
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-Smac/Diablo Antibody, Chemicon®, from rabbit