Merck
  • Protein secondary structure signatures from energy loss spectra recorded in the electron microscope.
Protein secondary structure signatures from energy loss spectra recorded in the electron microscope.
Journal of microscopy (2020-12-12)
Katia March, Kartik Venkatraman, Chloe Du Truong, Dewight Williams, Po-Lin Chiu, Peter Rez
ABSTRACT
Infrared spectroscopy is a powerful technique for characterising protein structure. It is now possible to record energy losses corresponding to the infrared region in the electron microscope and to avoid damage by positioning the probe in the region adjacent to the structure being studied. Spectra from bacteriorhodopsin, a protein that is predominately a α helix, and OmpF porin, a protein that is mainly β sheet show significant differences over a spectral range from ∼0.1 to 0.25 eV (∼1000 to 1800 cm-1 ). Although the energy resolution equivalent to 60 cm-1 is inferior to Fourier Transform InfraRed Spectroscopy (FTIR) the spectra are very sensitive to molecular orientation. Polar bonds aligned parallel to the specimen grid make particularly strong contributions to the energy loss spectra. Ultra-high-resolution energy loss spectroscopy in the electron microscope can potentially add useful information to imaging and diffraction for determining the secondary structure misfolding believed to be responsible for dementia diseases such as Alzheimer's.
Sigma-Aldrich
Bacteriorhodopsin from Halobacterium salinarum, native sequence, lyophilized powder
Pricing and availability is not currently available.