Merck
  • Biotransformation of the triketone herbicide mesotrione by a Bacillus strain. Metabolite profiling using liquid chromatography/electrospray ionization quadrupole time-of-flight mass spectrometry.
Biotransformation of the triketone herbicide mesotrione by a Bacillus strain. Metabolite profiling using liquid chromatography/electrospray ionization quadrupole time-of-flight mass spectrometry.
Rapid communications in mass spectrometry : RCM (2006-08-01)
Stéphanie Durand, Bertrand Légeret, Anne-Sophie Martin, Martine Sancelme, Anne-Marie Delort, Pascale Besse-Hoggan, Bruno Combourieu
ABSTRACT
The metabolic pathway involved in the biotransformation of the herbicide mesotrione by the bacterial strain Bacillus sp. 3B6 was investigated by a reliable liquid chromatography/electrospray ionization quadrupole time-of-flight mass spectrometry (LC/ESI-QTOF-MS) method. The LC/ESI-MS method, both in positive and negative mode, with the assistance of MS(2) fragments and isotopic pattern analyses, allowed us to identify five metabolites. This work constitutes the first complete monitoring of mesotrione degradation kinetics. Among the transformation products found by both techniques, one was formed by intramolecular cyclization between a hydroxylamine and a keto function, which is quite a rare biological reactivity process. For each identified metabolite, a fragmentation pathway is proposed for negative and positive mode.
Product Description
Supelco
Mesotrion Metabolite AMBA, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland