Merck
  • Effects of cetrorelix, a GnRH-receptor antagonist, on gonadal axis in women with functional hypothalamic amenorrhea.
Effects of cetrorelix, a GnRH-receptor antagonist, on gonadal axis in women with functional hypothalamic amenorrhea.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2011-01-06)
Rita Berardelli, Laura Gianotti, Ioannis Karamouzis, Andreea Picu, Roberta Giordano, Valentina D'Angelo, Domenico Zinnà, Fabio Lanfranco, Ezio Ghigo, Emanuela Arvat
ABSTRACT
Gonadotropin Releasing Hormone (GnRH) antagonists (GnRHa) suppress gonadotropin and sex-steroid secretion. In normal women, acute GnRHa administration induces inhibitory effect on pituitary-gonadal axis, followed by Luteinizing Hormone (LH) rebound. Functional hypothalamic amenorrhea (HA) is characterised by impaired gonadotropin secretion and hypogonadism secondary to blunted GnRH pulsatility. We studied the effects of a GnRHa, cetrorelix (CTX 3.0 mg), in six women with HA (age 30.7 ± 3.2 years; BMI 21.5 ± 1.7 kg/m(2)) and six control subjects (CS, 28.2 ± 0.6 years; 22.6 ± 0.9 kg/m(2)) on LH, Follicle-Stimulating Hormone (FSH) and oestradiol levels over 4 h (08.00-12.00 am) before, +24 h and +96 h after CTX; LH, FSH, and oestradiol were also evaluated at +6, +8, +12, +48, +72 h after CTX. CS: CTX reduced (p < 0.05) LH, FSH, and oestradiol (nadir at +12 h, +24 h, and +24 h); LH rebounded at +96 h, FSH and oestradiol recovered at +48 h and +72 h. The 4-h evaluation showed LH and FSH reduction (p < 0.05) at +24 h, with LH rebound at +96 h. HA: CTX reduced (p < 0.05) LH, FSH, and oestradiol, (nadir at +24 h, +48 h, and +48 h, recovery at +48 h, +72 h, and +96 h). The 4-h evaluation showed gonadotropin reduction (p < 0.05) 24 h after CTX, without any rebound effect. One single CTX dose still modulates gonadotropin secretion in HA. Its 'paradoxical' stimulatory effect on gonadotropins needs to be verified after prolonged administration.