Merck
  • Specific viral RNA drives the SARS CoV-2 nucleocapsid to phase separate.
Specific viral RNA drives the SARS CoV-2 nucleocapsid to phase separate.
bioRxiv : the preprint server for biology (2020-06-27)
Christiane Iserman, Christine Roden, Mark Boerneke, Rachel Sealfon, Grace McLaughlin, Irwin Jungreis, Chris Park, Avinash Boppana, Ethan Fritch, Yixuan J Hou, Chandra Theesfeld, Olga G Troyanskaya, Ralph S Baric, Timothy P Sheahan, Kevin Weeks, Amy S Gladfelter
ABSTRACT
A mechanistic understanding of the SARS-CoV-2 viral replication cycle is essential to develop new therapies for the COVID-19 global health crisis. In this study, we show that the SARS-CoV-2 nucleocapsid protein (N-protein) undergoes liquid-liquid phase separation (LLPS) with the viral genome, and propose a model of viral packaging through LLPS. N-protein condenses with specific RNA sequences in the first 1000 nts (5'-End) under physiological conditions and is enhanced at human upper airway temperatures. N-protein condensates exclude non-packaged RNA sequences. We comprehensively map sites bound by N-protein in the 5'-End and find preferences for single-stranded RNA flanked by stable structured elements. Liquid-like N-protein condensates form in mammalian cells in a concentration-dependent manner and can be altered by small molecules. Condensation of N-protein is sequence and structure specific, sensitive to human body temperature, and manipulatable with small molecules thus presenting screenable processes for identifying antiviral compounds effective against SARS-CoV-2.
Sigma-Aldrich
(R)-(+)-α-Lipoic acid, ≥98.0% (HPLC)
Sigma-Aldrich
1,6-Hexanediol, 99%