Merck
  • Interplay between chromatin remodeling and epigenetic changes during lineage-specific commitment to granzyme B expression.
Interplay between chromatin remodeling and epigenetic changes during lineage-specific commitment to granzyme B expression.
Journal of immunology (Baltimore, Md. : 1950) (2009-11-17)
Torsten Juelich, Elissa L Sutcliffe, Elissa Sutcliffe, Alice Denton, Yiqing He, Peter C Doherty, Christopher R Parish, Christopher Parish, Stephen J Turner, Steven J Turner, David J Tremethick, David Tremethick, Sudha Rao
ABSTRACT
The role of chromatin remodeling and histone posttranslational modifications and how they are integrated to control gene expression during the acquisition of cell-specific functions is poorly understood. We show here that following in vitro activation of CD4(+) and CD8(+) T lymphocytes, both cell types show rapid histone H3 loss at the granzyme B (gzmB) proximal promoter region. However, despite the gzmB proximal promoter being remodeled in both T cell subsets, only CD8(+) T cells express high levels of gzmB and display a distinct pattern of key epigenetic marks, notably differential H3 acetylation and methylation. These data suggest that for high levels of transcription to occur a distinct set of histone modifications needs to be established in addition to histone loss at the proximal promoter of gzmB.