Merck
  • Inhibition of acute leukemia with attenuated Salmonella typhimurium strain VNP20009.
Inhibition of acute leukemia with attenuated Salmonella typhimurium strain VNP20009.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-06-23)
Meirong Li, Mengmeng Lu, Yunhao Lai, Xindan Zhang, Yuyu Li, Ping Mao, Zhicheng Liang, Yunping Mu, Ying Lin, Allan Z Zhao, Zhenggang Zhao, Sujin Zhou, Fanghong Li
ABSTRACT
Acute leukemia is a common hematological malignancy. Despite recent promising progress, the prognosis of acute leukemia patients remains to be improved. New therapies are therefore still needed. Salmonella typhimurium has been shown to be highly effective as an anti-tumor agent in many solid cancer models, but it has not been applied in acute leukemia. Here, we report an attenuated Salmonella typhimurium strain, VNP20009, can induce apoptosis in multiple types of leukemia cells both in vivo and in vitro. Furthermore, VNP20009 significantly inhibited the proliferation of MLL-AF9-induced acute myeloid leukemia cells and prolonged the survival of the AML-carrying mice. VNP20009 restored the counts of white blood cell (WBC) and its five subsets in peripheral blood (PB) to near-physiological values, and elevated the levels of certain cytokines, such as tumor necrosis factor-α (TNF-α), leukemia inhibitory factor (LIF), interferon-γ (IFN-γ), chemokine C-X-C motif ligand-10 (CXCL-10) and C-C motif ligand-2 (CCL-2). Moreover, the ratio of immune cells, including natural killer cells (NKs), CD4+ Th1-type cells and CD8+ IFN-γ-producing effector T cells were highly upregulated in the AML mice treated with VNP20009. The results of the present study potentially provide an alternative therapeutic strategy for hematologic malignancies through boosting the innate and adaptive anti-tumor immunity.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
(+)-Brefeldin A, Eupenicillium brefeldianum, Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid