Merck
  • Sushi domain-containing protein 4 controls synaptic plasticity and motor learning.
Sushi domain-containing protein 4 controls synaptic plasticity and motor learning.
eLife (2021-03-05)
Inés González-Calvo, Keerthana Iyer, Mélanie Carquin, Anouar Khayachi, Fernando A Giuliani, Séverine M Sigoillot, Jean Vincent, Martial Séveno, Maxime Veleanu, Sylvana Tahraoui, Mélanie Albert, Oana Vigy, Célia Bosso-Lefèvre, Yann Nadjar, Andréa Dumoulin, Antoine Triller, Jean-Louis Bessereau, Laure Rondi-Reig, Philippe Isope, Fekrije Selimi
ABSTRACT
Fine control of protein stoichiometry at synapses underlies brain function and plasticity. How proteostasis is controlled independently for each type of synaptic protein in a synapse-specific and activity-dependent manner remains unclear. Here, we show that Susd4, a gene coding for a complement-related transmembrane protein, is expressed by many neuronal populations starting at the time of synapse formation. Constitutive loss-of-function of Susd4 in the mouse impairs motor coordination adaptation and learning, prevents long-term depression at cerebellar synapses, and leads to misregulation of activity-dependent AMPA receptor subunit GluA2 degradation. We identified several proteins with known roles in the regulation of AMPA receptor turnover, in particular ubiquitin ligases of the NEDD4 subfamily, as SUSD4 binding partners. Our findings shed light on the potential role of SUSD4 mutations in neurodevelopmental diseases.
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Anti-Nedd4, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor Delta 1/2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Roche
NBT/BCIP Stock Solution