Merck
  • Polyclonal F(ab')2 fragments of equine antibodies raised against the spike protein neutralize SARS-CoV-2 variants with high potency.
Polyclonal F(ab')2 fragments of equine antibodies raised against the spike protein neutralize SARS-CoV-2 variants with high potency.
iScience (2021-11-02)
Luis Eduardo R Cunha, Adilson A Stolet, Marcelo A Strauch, Victor A R Pereira, Carlos H Dumard, Andre M O Gomes, Fábio L Monteiro, Luiza M Higa, Patrícia N C Souza, Juliana G Fonseca, Francisco E Pontes, Leonardo G R Meirelles, José W M Albuquerque, Carolina Q Sacramento, Natalia Fintelman-Rodrigues, Tulio M Lima, Renata G F Alvim, Federico F Marsili, Marcella Moreira Caldeira, Russolina B Zingali, Guilherme A P de Oliveira, Thiago M L Souza, Alexandre S Silva, Rodrigo Muller, Daniela Del Rosário Flores Rodrigues, Luciana Jesus da Costa, Arthur Daniel R Alves, Marcelo Alves Pinto, Andréa C Oliveira, Herbert L M Guedes, Amilcar Tanuri, Leda R Castilho, Jerson L Silva
ABSTRACT
We used the recombinant trimeric spike (S) glycoprotein in the prefusion conformation to immunize horses for the production of hyperimmune globulins against SARS-CoV-2. Serum antibody titers measured by ELISA were above 1:106, and the neutralizing antibody titer against authentic virus (WT) was 1:14,604 (average PRNT90). Plasma from immunized animals was pepsin digested to remove the Fc portion and purified, yielding an F(ab')2 preparation with PRNT90 titers 150-fold higher than the neutralizing titers in human convalescent plasma. Challenge studies were carried out in hamsters and showed the in vivo ability of equine F(ab')2 to reduce viral load in the pulmonary tissues and significant clinical improvement determined by weight gain. The neutralization curve by F(ab')2 was similar against the WT and P.2 variants, but displaced to higher concentrations by 0.39 log units against the P.1 (Gamma) variant. These results support the possibility of using equine F(ab')2 preparation for the clinical treatment of COVID patients.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Horse IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution