Merck
  • A capillary-based microfluidic device enables primary high-throughput room-temperature crystallographic screening.
A capillary-based microfluidic device enables primary high-throughput room-temperature crystallographic screening.
Journal of applied crystallography (2021-08-26)
Shuo Sui, Anne Mulichak, Raviraj Kulathila, Joshua McGee, Danny Filiatreault, Sarthak Saha, Aina Cohen, Jinhu Song, Holly Hung, Jonathan Selway, Christina Kirby, Om K Shrestha, Wilhelm Weihofen, Michelle Fodor, Mei Xu, Rajiv Chopra, Sarah L Perry
ABSTRACT
A novel capillary-based microfluidic strategy to accelerate the process of small-molecule-compound screening by room-temperature X-ray crystallography using protein crystals is reported. The ultra-thin microfluidic devices are composed of a UV-curable polymer, patterned by cleanroom photolithography, and have nine capillary channels per chip. The chip was designed for ease of sample manipulation, sample stability and minimal X-ray background. 3D-printed frames and cassettes conforming to SBS standards are used to house the capillary chips, providing additional mechanical stability and compatibility with automated liquid- and sample-handling robotics. These devices enable an innovative in situ crystal-soaking screening workflow, akin to high-throughput compound screening, such that quantitative electron density maps sufficient to determine weak binding events are efficiently obtained. This work paves the way for adopting a room-temperature microfluidics-based sample delivery method at synchrotron sources to facilitate high-throughput protein-crystallography-based screening of compounds at high concentration with the aim of discovering novel binding events in an automated manner.
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications