Merck
DNA looping by protamine follows a nonuniform spatial distribution.
Biophysical journal (2021-05-24)
Ryan B McMillan, Victoria D Kuntz, Luka M Devenica, Hilary Bediako, Ashley R Carter
ABSTRACT
DNA looping plays an important role in cells in both regulating and protecting the genome. Often, studies of looping focus on looping by prokaryotic transcription factors like lac repressor or by structural maintenance of chromosomes proteins such as condensin. Here, however, we are interested in a different looping method whereby condensing agents (charge ≥+3) such as protamine proteins neutralize the DNA, causing it to form loops and toroids. We considered two previously proposed mechanisms for DNA looping by protamine. In the first mechanism, protamine stabilizes spontaneous DNA fluctuations, forming randomly distributed loops along the DNA. In the second mechanism, protamine binds and bends the DNA to form a loop, creating a distribution of loops that is biased by protamine binding. To differentiate between these mechanisms, we imaged both spontaneous and protamine-induced loops on short-length (≤1 μm) DNA fragments using atomic force microscopy. We then compared the spatial distribution of the loops to several model distributions. A random looping model, which describes the mechanism of spontaneous DNA folding, fit the distribution of spontaneous loops, but it did not fit the distribution of protamine-induced loops. Specifically, it failed to predict a peak in the spatial distribution of loops at an intermediate location along the DNA. An electrostatic multibinding model, which was created to mimic the bind-and-bend mechanism of protamine, was a better fit of the distribution of protamine-induced loops. In this model, multiple protamines bind to the DNA electrostatically within a particular region along the DNA to coordinate the formation of a loop. We speculate that these findings will impact our understanding of protamine's in vivo role for looping DNA into toroids and the mechanism of DNA condensation by condensing agents more broadly.
Sigma-Aldrich
Protamine from salmon, Grade IV, Histone, free(Millon test)