Merck
  • Synthesis, biological evaluation and mechanistic studies of totarol amino alcohol derivatives as potential antimalarial agents.
Synthesis, biological evaluation and mechanistic studies of totarol amino alcohol derivatives as potential antimalarial agents.
Bioorganic & medicinal chemistry (2011-12-27)
Claire Tacon, Eric M Guantai, Peter J Smith, Kelly Chibale
ABSTRACT
Herein we report on the semisynthesis and biological evaluation of β-amino alcohol derivatives of the natural product totarol and other simple aromatic systems. All β-amino alcohol derivatives of totarol exhibited higher antiplasmodial activity than totarol [IC(50): 11.69 μM (K1, chloroquine and multi-drug resistant strain), and 11.78 μM (D10, chloroquine sensitive strain)]-12e was the most active [IC(50): 0.63 μM (K1), and 0.61 μM (D10)]. The phenyl and naphthyl β-amino alcohol derivatives were much less active than their corresponding totarol equivalents. The majority of the β-amino alcohol derivatives of totarol were more active against K1 than the D10 strains of Plasmodium falciparum, a trend similar to the inverse relationship observed with the established aryl-amino alcohol antimalarial mefloquine. Selected compounds were shown to affect erythrocyte morphology, inhibit erythrocyte invasion and trigger CQ accumulation.
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
1,2-Epoxy-3-phenoxypropane, 99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1,2-Epoxy-3-phenoxypropane, technical grade
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)