Merck
A Contaminant Impurity, Not Rigosertib, Is a Tubulin Binding Agent.
Molecular cell (2020-07-04)
Stacey J Baker, Stephen C Cosenza, Saikrishna Athuluri-Divakar, M V Ramana Reddy, Rodrigo Vasquez-Del Carpio, Rinku Jain, Aneel K Aggarwal, E Premkumar Reddy
ABSTRACT
Rigosertib is a styryl benzyl sulfone that inhibits growth of tumor cells and acts as a RAS mimetic by binding to Ras binding domains of RAS effectors. A recent study attributed rigosertib's mechanism of action to microtubule binding. In that study, rigosertib was obtained from a commercial vendor. We compared the purity of clinical-grade and commercially sourced rigosertib and found that commercially sourced rigosertib contains approximately 5% ON01500, a potent inhibitor of tubulin polymerization. Clinical-grade rigosertib, which is free of this impurity, does not exhibit tubulin-binding activity. Cell lines expressing mutant β-tubulin have also been reported to be resistant to rigosertib. However, our study showed that these cells failed to proliferate in the presence of rigosertib at concentrations that are lethal to wild-type cells. Rigosertib induced a senescence-like phenotype in the small percentage of surviving cells, which could be incorrectly scored as resistant using short-term cultures.
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Pricing and availability is not currently available.