Merck
Neuronal non-CG methylation is an essential target for MeCP2 function.
Molecular cell (2021-02-10)
Rebekah Tillotson, Justyna Cholewa-Waclaw, Kashyap Chhatbar, John C Connelly, Sophie A Kirschner, Shaun Webb, Martha V Koerner, Jim Selfridge, David A Kelly, Dina De Sousa, Kyla Brown, Matthew J Lyst, Skirmantas Kriaucionis, Adrian Bird
ABSTRACT
DNA methylation is implicated in neuronal biology via the protein MeCP2, the mutation of which causes Rett syndrome. MeCP2 recruits the NCOR1/2 co-repressor complexes to methylated cytosine in the CG dinucleotide, but also to sites of non-CG methylation, which are abundant in neurons. To test the biological significance of the dual-binding specificity of MeCP2, we replaced its DNA binding domain with an orthologous domain from MBD2, which can only bind mCG motifs. Knockin mice expressing the domain-swap protein displayed severe Rett-syndrome-like phenotypes, indicating that normal brain function requires the interaction of MeCP2 with sites of non-CG methylation, specifically mCAC. The results support the notion that the delayed onset of Rett syndrome is due to the simultaneous post-natal accumulation of mCAC and its reader MeCP2. Intriguingly, genes dysregulated in both Mecp2 null and domain-swap mice are implicated in other neurological disorders, potentially highlighting targets of relevance to the Rett syndrome phenotype.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-MeCP2 antibody, Mouse monoclonal, clone Mec-168, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MeCP2 antibody, Mouse monoclonal, clone Men-8, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-HDAC3 antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Cy3 Conjugate, clone A60, from mouse, CY3 conjugate