Merck
  • Discovery of SHP2-D26 as a First, Potent, and Effective PROTAC Degrader of SHP2 Protein.
Discovery of SHP2-D26 as a First, Potent, and Effective PROTAC Degrader of SHP2 Protein.
Journal of medicinal chemistry (2020-05-22)
Mingliang Wang, Jianfeng Lu, Mi Wang, Chao-Yie Yang, Shaomeng Wang
ABSTRACT
Src homology 2 domain-containing phosphatase 2 (SHP2) is an attractive therapeutic target for human cancers and other human diseases. Herein, we report our discovery of potent small-molecule SHP2 degraders whose design is based upon the proteolysis-targeting chimera (PROTAC) concept. This work has led to the discovery of potent and effective SHP2 degraders, exemplified by SHP2-D26. SHP2-D26 achieves DC50 values of 6.0 and 2.6 nM in esophageal cancer KYSE520 and acute myeloid leukemia MV4;11 cells, respectively, and is capable of reducing SHP2 protein levels by >95% in cancer cells. SHP2-D26 is >30-times more potent in inhibition of phosphorylation of extracellular signal-regulated kinase (ERK) and of cell growth than SHP099, a potent SHP2 inhibitor, in KYSE520 and MV4;11 cancer cell lines. This study demonstrates that induced SHP2 degradation is a very effective approach to inhibit the function of SHP2. Further optimization of these SHP2 degraders may lead to the development of a new class of therapies for cancers and other human diseases.
Sigma-Aldrich
(S,R,S)-AHPC-Me, 95%