Merck
  • N-acylaminophenothiazines: neuroprotective agents displaying multifunctional activities for a potential treatment of Alzheimer's disease.
N-acylaminophenothiazines: neuroprotective agents displaying multifunctional activities for a potential treatment of Alzheimer's disease.
European journal of medicinal chemistry (2011-03-23)
Gema C González-Muñoz, Mariana P Arce, Beatriz López, Concepción Pérez, Alejandro Romero, Laura del Barrio, María Dolores Martín-de-Saavedra, Javier Egea, Rafael León, Mercedes Villarroya, Manuela G López, Antonio G García, Santiago Conde, María Isabel Rodríguez-Franco
ABSTRACT
We have previously reported the multifunctional profile of N-(3-chloro-10H-phenothiazin-10-yl)-3-(dimethylamino)propanamide (1) as an effective neuroprotectant and selective butyrylcholinesterase inhibitor. In this paper, we have developed a series of N-acylaminophenothiazines obtained from our compound library or newly synthesised. At micro- and sub-micromolar concentrations, these compounds selectively inhibited butyrylcholinesterase (BuChE), protected neurons against damage caused by both exogenous and mitochondrial free radicals, showed low toxicity, and could penetrate into the CNS. In addition, N-(3-chloro-10H-phenothiazin-10-yl)-2-(pyrrolidin-1-yl)acetamide (11) modulated the cytosolic calcium concentration and protected human neuroblastoma cells against several toxics, such as calcium overload induced by an L-type Ca2+-channel agonist, tau-hyperphosphorylation induced by okadaic acid and Aβ peptide.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Ofloxacin, VETRANAL®, analytical standard