Merck
  • Novel ambler class A carbapenem-hydrolyzing beta-lactamase from a Pseudomonas fluorescens isolate from the Seine River, Paris, France.
Novel ambler class A carbapenem-hydrolyzing beta-lactamase from a Pseudomonas fluorescens isolate from the Seine River, Paris, France.
Antimicrobial agents and chemotherapy (2009-11-11)
Delphine Girlich, Laurent Poirel, Patrice Nordmann
ABSTRACT
A Pseudomonas fluorescens isolate (PF-1) resistant to carbapenems was recovered during an environmental survey performed with water from the Seine River (Paris). It expressed a novel Ambler class A carbapenemase, BIC-1, sharing 68 and 59% amino acid identities with beta-lactamases SFC-1 from Serratia fonticola and the plasmid-encoded KPC-2, respectively. beta-Lactamase BIC-1 hydrolyzed penicillins, carbapenems, and cephalosporins except ceftazidime and monobactams. The bla(BIC-1) gene was chromosomally located and was also identified in two other P. fluorescens strains isolated from the Seine River 3 months later.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis