Merck
  • Analysis of physico-chemical properties of substrates of ABC and MFS multidrug transporters of pathogenic Candida albicans.
Analysis of physico-chemical properties of substrates of ABC and MFS multidrug transporters of pathogenic Candida albicans.
European journal of medicinal chemistry (2010-08-27)
Nidhi Puri, Om Prakash, Raman Manoharlal, Monika Sharma, Indira Ghosh, Rajendra Prasad
ABSTRACT
In this study, we have explored the structure activity relationships of substrates of two major, promiscuous, multidrug transporters of an opportunistic human pathogen Candida albicans namely, CaCdr1p and CaMdr1p. To differentiate between substrates and non-substrates, the susceptibilities of the Saccharomyces cerevisiae strains over-expressing CaCdr1p or CaMdr1p were determined for 67 structurally diverse xenobiotics. A comparison of physico-chemical indices of these tested compounds enabled identification of molecular descriptors such as, degree of hydrophobicity (MLogP), geometrical descriptor (DISPv), molecular edge descriptor (MDEC.12 and MDEC.13) and 3D-Morse descriptors, that allowed their segregation into substrates and non-substrates for both the transporter proteins. Taken together, present study provides first evidence of chemical basis of substrate specificities of two clinically relevant multidrug transporters of an opportunistic human pathogen C. albicans.
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, 95%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
6-Benzylaminopurine solution, 1 mg/mL, suitable for plant cell culture
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)