Merck
  • Solvent molecules form surface redox mediators in situ and cocatalyze O2 reduction on Pd.
Solvent molecules form surface redox mediators in situ and cocatalyze O2 reduction on Pd.
Science (New York, N.Y.) (2021-02-06)
Jason S Adams, Ashwin Chemburkar, Pranjali Priyadarshini, Tomas Ricciardulli, Yubing Lu, Vineet Maliekkal, Abinaya Sampath, Stuart Winikoff, Ayman M Karim, Matthew Neurock, David W Flaherty
ABSTRACT
Solvent molecules influence the reactions of molecular hydrogen and oxygen on palladium nanoparticles. Organic solvents activate to form reactive surface intermediates that mediate oxygen reduction through pathways distinct from reactions in pure water. Kinetic measurements and ab initio quantum chemical calculations indicate that methanol and water cocatalyze oxygen reduction by facilitating proton-electron transfer reactions. Methanol generates hydroxymethyl intermediates on palladium surfaces that efficiently transfer protons and electrons to oxygen to form hydrogen peroxide and formaldehyde. Formaldehyde subsequently oxidizes hydrogen to regenerate hydroxymethyl. Water, on the other hand, heterolytically oxidizes hydrogen to produce hydronium ions and electrons that reduce oxygen. These findings suggest that reactions of solvent molecules at solid-liquid interfaces can generate redox mediators in situ and provide opportunities to substantially increase rates and selectivities for catalytic reactions.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Palladium(II) nitrate dihydrate, ~40% Pd basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-d3, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-OD, 99 atom % D
Pricing and availability is not currently available.