Merck
  • Mapping Physiological ADP-Ribosylation Using Activated Ion Electron Transfer Dissociation.
Mapping Physiological ADP-Ribosylation Using Activated Ion Electron Transfer Dissociation.
Cell reports (2020-09-24)
Sara C Buch-Larsen, Ivo A Hendriks, Jean M Lodge, Martin Rykær, Benjamin Furtwängler, Evgenia Shishkova, Michael S Westphall, Joshua J Coon, Michael L Nielsen
ABSTRACT
ADP-ribosylation (ADPr) is a post-translational modification that plays pivotal roles in a wide range of cellular processes. Mass spectrometry (MS)-based analysis of ADPr under physiological conditions, without relying on genetic or chemical perturbation, has been hindered by technical limitations. Here, we describe the applicability of activated ion electron transfer dissociation (AI-ETD) for MS-based proteomics analysis of physiological ADPr using our unbiased Af1521 enrichment strategy. To benchmark AI-ETD, we profile 9,000 ADPr peptides mapping to >5,000 unique ADPr sites from a limited number of cells exposed to oxidative stress and identify 120% and 28% more ADPr peptides compared to contemporary strategies using ETD and electron-transfer higher-energy collisional dissociation (EThcD), respectively. Under physiological conditions, AI-ETD identifies 450 ADPr sites on low-abundant proteins, including in vivo cysteine modifications on poly(ADP-ribosyl)polymerase (PARP) 8 and tyrosine modifications on PARP14, hinting at specialist enzymatic functions for these enzymes. Collectively, our data provide insights into the physiological regulation of ADPr.
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
PARG human, recombinant, expressed in Sf21 cells, His tagged, >95% (SDS-PAGE)
Pricing and availability is not currently available.