Merck
  • Identification of signature genes associated with therapeutic resistance to anti-VEGF therapy.
Identification of signature genes associated with therapeutic resistance to anti-VEGF therapy.
Oncotarget (2020-02-01)
Pharavee Jaiprasart, Samrita Dogra, Deepika Neelakantan, Bharat Devapatla, Sukyung Woo
ABSTRACT
VEGF-mediated tumor angiogenesis is a validated clinical target in many cancers, but modest efficacy and rapid development of resistance are major challenges of VEGF-targeted therapies. To establish a molecular signature of this resistance in ovarian cancer, we developed preclinical tumor models of adaptive resistance to chronic anti-VEGF treatment. We performed RNA-seq analysis and reverse-phase protein array to compare changes in gene and protein expressions in stroma and cancer cells from resistant and responsive tumors. We identified a unique set of stromal-specific genes that were strongly correlated with resistance phenotypes against two different anti-VEGF treatments, and selected the apelin/APJ signaling pathway for further in vitro validation. Using various functional assays, we showed that activation of apelin/APJ signaling reduces the efficacy of a VEGF inhibitor in endothelial cells. In patients with ovarian cancer treated with bevacizumab, increased expression of apelin was associated with significantly decreased disease-free survival. These findings link signature gene expressions with anti-VEGF response, and may thus provide novel targetable mechanisms of clinical resistance to anti-VEGF therapies.
Sigma-Aldrich
SU1498, ≥98% (HPLC)