Merck
  • Protocol for Quantitative Analysis of Synaptic Vesicle Clustering in Axons of Cultured Neurons.
Protocol for Quantitative Analysis of Synaptic Vesicle Clustering in Axons of Cultured Neurons.
STAR protocols (2020-10-29)
Ji Won Um, Kyung Ah Han, Se-Young Choi, Jaewon Ko
ABSTRACT
Clustering of synaptic vesicles along the neuronal axons is a critical mechanism underpinning proper synaptic transmission. Here, we provide a detailed protocol for analyzing the distribution of synaptic vesicles in presynaptic boutons of cultured neurons. The protocol covers preparation of cultured neurons, expression of synaptic vesicle-enriched proteins, and quantification procedures. Utilizing neurons from postnatal transgenic mice, this method can be applied to investigate the roles of synaptic genes in regulating vesicle dynamics at synaptic sites. For complete details on the use and execution of this protocol, please refer to Han et al. (2020a).
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Triton X-100, for molecular biology