Merck
  • Effect of follicle size on oocytes recovery rate, quality, and in-vitro developmental competence in Bos indicus cows.
Effect of follicle size on oocytes recovery rate, quality, and in-vitro developmental competence in Bos indicus cows.
Animal reproduction (2020-10-09)
Zaeem Sarwar, Muhammad Saad, Muhammad Saleem, Ali Husnain, Amjad Riaz, Nasim Ahmad
ABSTRACT
The objective of the present study was to determine the effect of follicle size on recovery rate, quality, and in-vitro developmental competence of oocytes in Bos indicus cows. The ovaries (n = 507) of Bos indicus cows having age of 5-8 years, with mixed parity, BCS 2.75 ± 0.25, and clinically normal reproductive tracts were collected from the local abattoir. The follicles on the ovaries were divided into two groups based upon their size; 1) ≥6 mm diameter, and 2) <6 mm diameter. After initial evaluation of quality of the oocytes, the COCs were in vitro matured, fertilized, and cultured to determine the in vitro developmental competence. The oocyte recovery, quality, maturation, cleavage, 4-cell, 8-cell, and 16-cell stages were analyzed using PROC GLIMMIX procedure of SAS. However, the number of oocytes recovered per ovary was analyzed using MIXED procedure of SAS. Results revealed that the recovery of oocytes (LSM ± SEM) derived from the follicles having size <6 mm per ovary was greater (1.02 vs. 3.14 ± 0.13; P < 0. 0001). However, the percentage (n/n) recovery [69.8 (474/679) vs. 62.7% (1454/2320); P = 0.01] and grade I_+_II oocytes [68.4 (324/474) vs. 57.9% (842/1454); P < 0.0001] was greater in ≥6 mm as compared with <6 mm group, respectively. However, maturation rate did not differ [92.9 (288/310) vs. 92.2% (296/321); P = 0.98] between the groups. In contrast, cleavage rate [58.1 (180/310) vs. 47.4% (152/321); P = 0.01], the 4-cell [34.5 (107/310) vs. 18.7% (60/321); P = 0.0003], 8-cell [15.5 (48/310) vs. 7.8% (25/321); P = 0.008], and 16-cell [8.7 (27/310) vs. 2.1% (7/321); P = 0.004] stage embryos were greater in ≥6 mm group. It can be concluded that oocytes derived from follicle ≥6 mm have better in vitro developmental competence based on embryonic conversion in Bos indicus cows.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
MEM Amino Acids (50x) solution, Without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture