Merck
  • Functional characterization of SLC26A3 c.392C>G (p.P131R) mutation in intestinal barrier function using CRISPR/CAS9-created cell models.
Functional characterization of SLC26A3 c.392C>G (p.P131R) mutation in intestinal barrier function using CRISPR/CAS9-created cell models.
Cell & bioscience (2019-05-23)
Nini Zhang, Daniel P Heruth, Weibin Wu, Li Qin Zhang, Marianne N Nsumu, Katherine Shortt, Kelvin Li, Xun Jiang, Baoxi Wang, Craig Friesen, Ding-You Li, Shui Qing Ye
ABSTRACT
Congenital chloride diarrhea (CCD) in a newborn is a rare autosomal recessive disorder with life-threatening complications, requiring early diagnostics and treatment to prevent severe dehydration and infant mortality. SLC26A3 rs386833481 (c.392C>G; p.P131R) gene polymorphism is an important genetic determinant of CCD. Here, we report the influence of the non-synonymous SLC26A3 variant rs386833481 gene polymorphism on the function of the epithelial barrier and the potential mechanisms of these effects. We found that P131R-SLC26A3 increased dysfunction of the epithelial barrier compared with wild type SLC26A3 in human colonic Caco-2 and mouse colonic CMT-93 cells. When P131R-SLC26A3 was subsequently reverted to wild type, the epithelial barrier function was restored similar to wild type cells. Further study demonstrated that variant P131R-SLC26A3 disrupts function of epithelial barrier through two distinct molecular mechanisms: (a) decreasing SLC26A3 expression through a ubiquitination pathway and (b) disrupting a key interaction with its partner ZO-1/CFTR, thereby increasing the epithelial permeability. Our study provides an important insight of SLC26A3 SNPs in the regulation of the epithelial permeability and indicates that SLC26A3 rs386833481 is likely a causative mutation in the dysfunction of epithelial barrier of CCD, and correction of this SNP or increasing SLC26A3 function could be therapeutically beneficial for chronic diarrhea diseases.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.