Merck
  • Repurposing antitussive benproperine phosphate against pancreatic cancer depends on autophagy arrest.
Repurposing antitussive benproperine phosphate against pancreatic cancer depends on autophagy arrest.
Molecular oncology (2020-11-24)
Huanyu Zhang, Zhe Zhang, Yonghao Huang, Siyuan Qin, Li Zhou, Ningna Weng, Jiayang Liu, Mei Yang, Xiaodian Zhang, Yanda Lu, Lin Ma, Shaojiang Zheng, Qifu Li
ABSTRACT
Pancreatic cancer (PC) is one of the most common human malignancies worldwide and remains a major clinical challenge. Here, we found that benproperine phosphate (BPP), a cough suppressant, showed a significant anticancer effect on PC both in vitro and in vivo via the induction of autophagy-mediated cell death. Mechanistic studies revealed that BPP triggered AMPK/mTOR-mediated autophagy initiation and disturbed Ras-related protein Rab-11A (RAB11A)-mediated autophagosome-lysosome fusion, resulting in excessive accumulation of autophagosomes. Inhibition of autophagy or overexpression of RAB11A partially reversed BPP-induced growth inhibition in PC cells, suggesting that BPP might induce lethal autophagy arrest in PC cells. In conclusion, our results identify BPP as a potent antitumor agent for PC via the induction of autophagy arrest, therefore providing a new potential therapeutic strategy for the treatment of PC.
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acridine Orange hemi(zinc chloride) salt, For nucleic acid staining in cells or gels
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Anti-KI67 antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM360, purified immunoglobulin